Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHMT1-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024341.1 | 1 | 122 | |||
GenBank mRNA | AK097611.1 | - | 1 | 122 | ||
AK127990.1 | - | 1 | 122 |