Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242336.1 | NP_001229265.1 | 1 | 145 | ||
GenBank mRNA | AL037715.1 | - | 1 | 114 | ||
BC026244.1 | - | 1 | 105 | 4697443 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG531421.1 | - | 1 | 106 | Chr. 5 - 154038999 - 154050019 [+] on Build GRCh38 |