Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027156.1 | 4 | 1385 | |||
GenBank mRNA | AK308459.1 | - | 4 | 1385 |
Gene Symbol: TUBB8P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC139719.1 | - | 3 | 419 |