Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCT6P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003110.2 | 1, 2 | 173 | |||
GenBank mRNA | BC052238.1 | - | 1, 2 | 173 | ||
BC073761.1 | - | 1, 2 | 173 |