Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNTL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033169.1 | 5 | 756 | |||
XM_011515852.2 | XP_011514154.1 | 3 | 330 | |||
XM_011515853.2 | XP_011514155.1 | 2 | 88 | |||
GenBank mRNA | AK093091.1 | - | 5 | 709 | ||
AK315667.1 | - | 5 | 716 |