Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011520071.2 | XP_011518373.1 | 7 | 2723 |
Gene Symbol: CSNK2A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256686.1 | NP_001243615.1 | 1 | 28 |