Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC153684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015447.1 | 2 | 1989 | |||
GenBank mRNA | AK002146.1 | - | 3 | 951 | ||
AK123995.1 | - | 2 | 2031 | |||
BC101382.1 | - | 2 | 2041 | |||
BC101383.2 | - | 2 | 1733 | |||
BC101384.1 | - | 2 | 1990 | |||
BC101385.2 | - | 2 | 1989 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM555934.1 | - | 3 | 719 | Chr. 5 - 43041807 - 43045281 [+] on Build GRCh38 |