Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC153684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015447.1 | 2 | 1774 | |||
GenBank mRNA | AK002146.1 | - | 3 | 736 | ||
AK123995.1 | - | 2 | 1816 | |||
BC101382.1 | - | 2 | 1826 | |||
BC101383.2 | - | 2 | 1518 | |||
BC101384.1 | - | 2 | 1775 | |||
BC101385.2 | - | 2 | 1774 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM555934.1 | - | 3 | 504 | Chr. 5 - 43041807 - 43045281 [+] on Build GRCh38 |