Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PQLC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024016.2 | 3 | 318 | |||
GenBank mRNA | AA131625.1 | - | 3 | 294 | ||
BE905784.1 | - | 3 | 318 | |||
BX113701.1 | - | 3 | 289 |