Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4BP2L2-IT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026928.1 | 1 | 540 |
Gene Symbol: N4BP2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020342.1 | XP_016875831.1 | 7 | 6073 |