Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPG2IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024086.2 | 1 | 2331 | |||
GenBank mRNA | AF038190.1 | - | 1 | 773 | ||
AK123640.1 | - | 1 | 2326 |