Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015363.2 | 2 | 1417 | |||
NR_126044.1 | 2 | 1399 | ||||
NR_126045.1 | 3 | 675 | ||||
GenBank mRNA | AK092305.1 | - | 2 | 1316 | ||
AK095690.1 | - | 2 | 1300 | |||
BC000869.1 | - | 3 | 529 |
Gene Symbol: LOC102724238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126048.1 | 2 | 1343 |
Gene Symbol: LOC103908605 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126050.1 | 2 | 437 | |||
GenBank mRNA | DR003081.1 | - | 384 |
Gene Symbol: LOC554249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126049.1 | 2 | 1339 | |||
GenBank mRNA | BC084570.1 | - | 2 | 1253 | 6378300 |
Gene Symbol: LOC728673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126047.1 | 2 | 1301 | |||
GenBank mRNA | BC081570.1 | - | 1 | 638 |