Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015363.2 | 2 | 2305 | |||
NR_126044.1 | 2 | 2287 | ||||
NR_126045.1 | 3 | 1563 | ||||
GenBank mRNA | AK092305.1 | - | 2 | 2204 | ||
AK095690.1 | - | 2 | 2188 | |||
BC000869.1 | - | 3 | 1416 |
Gene Symbol: LOC728673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126047.1 | 2 | 2189 | |||
GenBank mRNA | BC081570.1 | - | 1 | 1526 |