Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102724238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126048.1 | 2 | 1054 |
Gene Symbol: GLIDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015363.2 | 2 | 1128 | |||
NR_126044.1 | 2 | 1110 | ||||
GenBank mRNA | AK092305.1 | - | 2 | 1027 | ||
AK095690.1 | - | 2 | 1011 | |||
BC027338.1 | - | 2 | 1014 |
Gene Symbol: LOC554249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126049.1 | 2 | 1050 | |||
GenBank mRNA | BC084570.1 | - | 2 | 964 | 6378300 |
Gene Symbol: LOC728673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126047.1 | 2 | 1012 | |||
GenBank mRNA | BC081570.1 | - | 1 | 349 |