Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC389765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029410.1 | 8 | 1543 | |||
GenBank mRNA | AK124523.1 | - | 5 | 1007 | ||
BC108718.1 | - | 8 | 1475 |