Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPHP3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002811.1 | 11 | 2451 | |||
GenBank mRNA | BC039375.1 | - | 11 | 2451 | ||
BC048124.1 | - | 11 | 2450 |