Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCBP2-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024388.1 | 1 | 766 | |||
GenBank mRNA | AK123020.1 | - | 1 | 717 | ||
AL832915.1 | - | 1 | 704 | |||
BC000257.1 | - | 1 | 700 | |||
BC007882.2 | - | 1 | 679 | |||
BC062368.1 | - | 1 | 766 |