Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RN7SK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_001445.2 | 1 | 298 | |||
GenBank mRNA | X04236.1 | - | LIT1704 7SK |
1 | 297 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK058127.1 | - | 298 | Chr. - - [-] on Build |