Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMNTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256266.1 | NP_001243195.1 | 2, 3 | 340 | ||
GenBank mRNA | BC037957.1 | - | 2, 3 | 309 | 5744078 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM563573.1 | - | 309 | Chr. - - [-] on Build |