Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRHDE-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026836.1 | 1 | 274 | |||
NR_026837.1 | 1 | 274 | ||||
GenBank mRNA | BC040230.1 | - | 1 | 249 | ||
DA024500.1 | - | 1 | 274 |