Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024357.2 | 2, 3 | 149 | |||
GenBank mRNA | AI793052.1 | - | 2, 3 | 149 |