Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR7E156P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002171.2 | 3, 4 | 2435 | |||
GenBank mRNA | AK097469.1 | - | 4, 5 | 2358 | ||
AK097490.1 | - | 3, 4 | 1674 | |||
AK097573.1 | - | 3, 4 | 1713 |