Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR7E156P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002171.2 | 4 | 2610 | |||
GenBank mRNA | AK097469.1 | - | 5 | 2533 | ||
AK097573.1 | - | 4 | 1888 |