Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORA33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002436.1 | 1 | 89 | |||
GenBank mRNA | AJ609473.1 | - | 1 | 89 | ||
AK096147.1 | - | 1 | 2736 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DW441862.1 | - | 1 | 105 | Chr. 6 - 132817219 - 132817439 [+] on Build GRCh38 |