Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYCAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003039.3 | 1 | 228 | |||
GenBank mRNA | AJ489589.1 | - | 1 | 195 | ||
AJ489590.1 | - | 1 | 195 | |||
AJ489592.1 | - | 1 | 228 | |||
AJ489593.1 | - | 1 | 228 |