Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGN2P46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_022014.1 | 2 | 144 | |||
GenBank mRNA | AK096745.1 | - | 3 | 221 | ||
AY271964.1 | - | 2 | 129 | |||
AY271965.1 | - | 2 | 129 | |||
AY271966.1 | - | 2 | 129 | |||
AY271967.1 | - | 2 | 129 |
Gene Symbol: ETV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EF632112.1 | - | 152 |