Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP83-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027035.1 | 1 | 1122 | |||
GenBank mRNA | AK057269.1 | - | 1 | 1122 | ||
BC035086.2 | - | 1 | 1105 | |||
BC044616.1 | - | 1 | 1106 |