Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLRN1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024066.1 | 6 | 1952 | |||
GenBank mRNA | AK126442.1 | - | 5 | 1234 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF388367.3 | - | 6 | 1952 | Chr. 3 - 150972678 - 151079830 [+] on Build GRCh38 |