Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024347.2 | 4 | 1988 | |||
GenBank mRNA | AK094020.1 | - | 4 | 1988 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL133082.1 | - | 1 | 1039 | Chr. 16 - 89162382 - 89164245 [+] on Build GRCh38 |