Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMCHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003921.1 | 3 | 355 | |||
GenBank mRNA | AY008407.1 | - | 4 | 433 | ||
AY008408.1 | - | 4 | 437 | |||
AY008409.1 | - | 3 | 355 | |||
AY008410.1 | - | 3 | 393 | |||
AY008411.1 | - | 3 | 1614 | |||
EU921424.1 | - | 4 | 269 | |||
EU921426.1 | - | 4 | 340 | |||
EU921427.1 | - | 4 | 380 | |||
EU921428.1 | - | 3 | 263 | |||
EU921429.1 | - | 4 | 340 | |||
EU921430.1 | - | 5 | 536 | |||
EU921435.1 | - | 3 | 214 | |||
EU938381.1 | - | 4 | 350 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX091674.1 | - | 3 | 281 | Chr. 5 - 22142464 - 22152246 [+] on Build GRCh38 |