Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMCHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003921.1 | 4 | 523 | |||
GenBank mRNA | AY008407.1 | - | 5 | 601 | ||
AY008408.1 | - | 5 | 605 | |||
AY008409.1 | - | 4 | 523 | |||
AY008410.1 | - | 4 | 561 | |||
AY008411.1 | - | 4 | 1782 | |||
EU921424.1 | - | 5 | 437 | |||
EU921426.1 | - | 5 | 508 | |||
EU921427.1 | - | 5 | 548 | |||
EU921428.1 | - | 4 | 431 | |||
EU921429.1 | - | 5 | 508 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX091674.1 | - | 4 | 449 | Chr. 5 - 22142464 - 22152246 [+] on Build GRCh38 |