Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027079.2 | 2 | 412 | |||
NR_027080.2 | 2 | 225 | ||||
GenBank mRNA | BC038774.1 | - | 2 | 225 | ||
BC045579.1 | - | 2 | 412 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI464475.1 | - | 2 | 230 | Chr. 16 - 35363412 - 35382466 [+] on Build GRCh38 |