Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CROT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243745.1 | NP_001230674.1 | 4 | 847 | ||
GenBank mRNA | BC006015.2 | - | 3 | 717 | ||
BC051874.1 | - | 4 | 789 |