Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029578.1 | 1 | 28 |
Gene Symbol: MIR194-2HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_133638.1 | 2 | 1988 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX505955.1 | - | 1 | 148 | Chr. 11 - 64890877 - 64891366 [-] on Build GRCh38 |