Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029627.1 | 1 | 66 |
Gene Symbol: DNM3OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038397.2 | 1 | 5994 | |||
NR_103486.1 | 4 | 2088 | ||||
GenBank mRNA | FJ623959.1 | - | 1 | 5994 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA439854.1 | - | 1 | 299 | Chr. 1 - 172138578 - 172139131 [-] on Build GRCh38 |