Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029513.1 | 1 | 33 | |||
GenBank mRNA | AF480536.1 | - | 1 | 12 | ||
AF480570.1 | - | 1 | 13 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA362525.1 | - | 1 | 42 | Chr. X - 53556072 - 53556350 [-] on Build GRCh38 |