Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4F35P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026756.1 | 1 | 1052 | |||
GenBank mRNA | BC037172.1 | - | 1 | 1052 |
Gene Symbol: CYP4F30P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093281.1 | - | 1 | 576 |
Gene Symbol: FAM95B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL833349.1 | - | 1 | 1472 |