Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4F35P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026756.1 | 4 | 2336 | |||
GenBank mRNA | BC037172.1 | - | 4 | 2336 |
Gene Symbol: ANKRD18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093059.1 | - | 8 | 1889 |
Gene Symbol: FAM95C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL834388.1 | - | 1 | 386 |