Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4F35P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026756.1 | 1 | 368 | |||
GenBank mRNA | BC037172.1 | - | 1 | 368 |
Gene Symbol: ANKRD18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093059.1 | - | 7 | 944 | ||
BC131497.1 | - | 7 | 1041 |
Gene Symbol: FAM95B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL833349.1 | - | 1 | 715 |
Gene Symbol: FAM95C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047651.1 | 2 | 1901 | |||
GenBank mRNA | AK097951.1 | - | 2 | 1903 |