Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLEU2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002771.1 | 1 | 1537 | |||
GenBank mRNA | AF254117.1 | - | 1 | 1537 |
Gene Symbol: DLEU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF264787.1 | - | 7 | 1899 |