Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLEU2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002771.1 | 1 | 31 | |||
GenBank mRNA | AF254117.1 | - | 1 | 31 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK022419.1 | - | 1 | 1929 | Chr. 1 - 63547082 - 63549558 [+] on Build GRCh38 |