Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026918.1 | 1 | 14 | |||
GenBank mRNA | AK303608.1 | - | 1 | 14 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DC410534.1 | - | 1 | 14 | Chr. 9 - 109406557 - 109417783 [-] on Build GRCh38 |