Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026918.1 | 1 | 93 | |||
GenBank mRNA | AK303608.1 | - | 1 | 93 | ||
AK308349.1 | - | 1 | 56 |