Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORA48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002918.1 | 1 | 67 | |||
GenBank mRNA | AJ609447.1 | - | 1 | 67 |
Gene Symbol: EIF4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U79273.1 | - | 1 | 34 |