Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CECR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015352.1 | 4 | 1954 | |||
GenBank mRNA | AK098045.1 | - | 4 | 1285 | ||
AY026052.1 | - | 4 | 1954 | |||
BC043198.2 | - | 4 | 303 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI600377.1 | - | 4 | 308 | Chr. 22 - 17037279 - 17058635 [+] on Build GRCh38 |