Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073199.1 | 3 | 219 | |||
GenBank mRNA | AK000670.1 | - | 3 | 220 | ||
AK000701.1 | - | 2 | 2154 | |||
AK310037.1 | - | 3 | 261 | |||
BP382918.1 | - | 3 | 219 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA925984.1 | - | 3 | 261 | Chr. 17 - 50763215 - 50767557 [-] on Build GRCh38 |