Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL23AP32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002229.2 | 1 | 402 | |||
GenBank mRNA | BC096706.1 | - | 1 | 402 | ||
BC096740.1 | - | 1 | 402 | |||
BC098172.1 | - | 1 | 402 |