Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030691.1 | 1 | 62 | |||
XM_006718602.2 | XP_006718665.1 | 1 | 58 | |||
XM_006718604.2 | XP_006718667.1 | 1 | 45 | |||
XM_006718605.2 | XP_006718668.1 | 1 | 57 | |||
XM_011545131.2 | XP_011543433.1 | 1 | 58 | |||
GenBank mRNA | AK097619.1 | - | 1 | 58 | ||
BC033036.2 | - | 1 | 18 | 4823645 | ||
DB445158.1 | - | 1 | 61 | |||
DB450993.1 | - | 1 | 50 |