Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT8-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024334.1 | 1 | 1542 | |||
GenBank mRNA | AK094893.1 | - | 1 | 1508 | ||
BC028147.1 | - | 1 | 1450 |