Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REG1CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002714.1 | 1 | 1838 | |||
GenBank mRNA | AK126561.1 | - | 1 | 1838 | ||
D56494.1 | - | 4 | 593 | |||
D56495.1 | - | 4 | 598 |